Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-S1PR4 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2883R-CY5.5
BOSSBS-2883R-CY5.5EA 900 EUR InStock
BOSSBS-2883R-CY5.5
Anti-S1PR4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
This protein belongs to a G protein coupled heptahelical receptor subfamily named Endothelial Cell Differentiation Genes (EDG)that act as receptors for biologically active lysophospholipids. This group consists of two receptor subgroups specific for S1P and LPA respectively. EDG6 is the receptor for lysophospholipid sphingosine 1 phosphate (S1P). S1P elicits diverse physiological effect on most types of cells and tissues. EDG6 may be involved in cell migration processes that are specific for lymphocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: S1PR4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge