Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IKZF3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2954R-A680
BOSSBS-2954R-A680EA 890 EUR InStock
BOSSBS-2954R-A680
Anti-IKZF3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
IKZF3 belongs to the Ikaros family of zinc-finger proteins. Members of this family are transcription factors that play an important role in the regulation of lymphocyte differentiation. They are involved in the control of gene expression and when associated with nuclear complexes, participate in nucleosome remodeling. IKZF3 is important in the regulation of B lymphocyte proliferation and differentiation.

Type: Primary
Antigen: IKZF3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/509
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge