Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PHGDH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2970R-HRP
BOSSBS-2970R-HRPEA 900 EUR InStock
BOSSBS-2970R-HRP
Anti-PHGDH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The survival and development of central neurons require the supply of trophic factors by glial cells. The trophic actions of glial cells on Purkinje neurons are mediated by L-serine and glycine, which are glia-derived trophic factors synthesized by 3PGDH (1). 3PGDH protein is 544 amino acids in length. Two distinct mRNA transcripts that encode for 3PGDH protein in normal human tissues are dominant 2.1 kb mRNA, which is highly expressed in prostate, testis, ovary, brain, liver, kidney, and pancreas, and weakly expressed in thymus, colon, and heart, and 710 bp mRNA, which is highly expressed in heart and skeletal muscle (2). 3PGDH is regulated at the transcriptional level depending on tissue specificity and cellular proliferative status (2). 3PGDH protein is also highly expressed in adult and fetal brain tissues (3). 3PGDH protein plays an important role in the metabolism, development, and function of the central nervous system (3) and its deficiency is a treatable congential error (4-5) that impairs L-serine biosynthesis which is characterized by congenital microcephaly, psychomotor retardation, and seizures (3).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PHGDH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge