Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CITED1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-2990R
BOSSBS-2990REA 725 EUR InStock
BOSSBS-2990R
Anti-CITED1 Rabbit Polyclonal Antibody
Antikörper
Transcriptional coactivator of the p300/CBP-mediated trancription complex. Enhances SMAD-mediated transcription by strengthening the functional link between the DNA-binding SMAD transcription factors and the p300/CBP transcription coactivator complex. Stimulates estrogen-dependent transactivation activity mediated by estrogen receptors signaling; stabilizes the interaction of estrogen receptor ESR1 and histone acetyltransferase EP300. Positively regulates TGF-beta signaling through its association with the SMAD/p300/CBP-mediated transcriptional coactivator complex. Induces transcription from estrogen-responsive promoters and protection against cell death. Potentiates EGR2-mediated transcriptional activation activity from the ERBB2 promoter. Acts as an inhibitor of osteoblastic mineralization through a cAMP-dependent parathyroid hormone receptor signaling. May play a role in pigmentation of melanocytes. Associates with chromatin to the estrogen-responsive TGF-alpha promoter region in a estrogen-dependent manner.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CITED1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge