Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MLLT4 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3072R
BOSSBS-3072REA 743 EUR InStock
BOSSBS-3072R
Anti-MLLT4 Rabbit Polyclonal Antibody
Antikörper
Afadin is a cell-cell adherens junction F-actin binding multidomain protein. Two splice variants of afadin have been described: l-afadin, which is ubiquitously expressed, and the smaller s-afadin, which is abundantly expressed in neural tissue. l-Afadin contains one PDZ domain near its middle followed by three proline-rich and one F-actin-binding region at the carboxyl-terminal. l/s-Afadin is localized to cell-cell junctions and binds several cytoplasmic proteins such as the small GTPase Ras, the tight junction protein ZO-1, and the vinculin binding protein ponsin/SH3P12.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MLLT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge