Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HDAC7 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3216R-CY3
BOSSBS-3216R-CY3EA 929 EUR InStock
BOSSBS-3216R-CY3
Anti-HDAC7 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
HDAC7 is a member of the class II mammalian histone deacetylases, which plays an important role in modulating the eukaryotic chromatin structure. Human HDAC7 is composed of 912 amino acid residues. Although HDAC7 is localized mostly to the cell nucleus, it is also found in the cytoplasm, suggesting nucleo-cytoplasmic shuttling. The histone deacetylase activity of HDAC7 maps to a carboxy-terminal domain and is dependent on interaction with class I HDACs in the nucleus. It is an active component of different transcriptional corepressor complexes that can be recruited to specific promoter regions via interactions with a growing number of sequence specific transcriptional factors. HDAC7 catalyzes removal of acetyl-groups from acetyl-lysines of histones and promotes compaction of chromatin in these regions, leading to the inhibition of gene transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HDAC7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge