Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PAR4 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3325R-CY5.5
BOSSBS-3325R-CY5.5EA 921 EUR InStock
BOSSBS-3325R-CY5.5
Anti-PAR4 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
PAR4 is a member of a unique family of GPCRs, the proteinase-activated receptors (PARs), that are activated by proteolytic cleavage of the N-terminal domain of the receptor to reveal a tethered ligand. The PAR family consists of 4 receptors; PAR1 and PAR3 are activated by thrombin, and PAR2 and PAR4 are activated by several serine proteases (Macfarlane et al., 2001). PAR4 is expressed along with PAR1 on human platelets, and each plays an important role in thrombin-induced platelet aggregation (Holinstat et al., 2006). However, platelets from mice express PAR4 but not PAR1, and platelets from mice lacking PAR4 fail to aggregate in response to thrombin (Sambrano et al., 2001). Mice lacking PAR4 are protected from mesenteric arteriole thrombosis, indicating that PAR4 is a potential target for treatment of thrombosis in humans.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAR4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge