Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PLCB3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3341R-CY5.5
BOSSBS-3341R-CY5.5EA 929 EUR InStock
BOSSBS-3341R-CY5.5
Anti-PLCB3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Phospholipase C is a key enzyme in phosphatidylinositol (PIP2) metabolism and lipid signaling pathways. The Phospholipase C family consists of 13 isozymes split between six subfamilies. Phospholipase C beta is activated by G protein subunits.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLCB3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge