Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DLG4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3349R-A488
BOSSBS-3349R-A488EA 921 EUR InStock
BOSSBS-3349R-A488
Anti-DLG4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
postsynaptic density protein 95 is a molecular scaffolding protein that binds and clusters N-methyl-D-aspartate receptors at neuronal synapses; may be involved in guanine nucleotide-mediated signal transduction pathway. This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with DLG2. With DLG2 it is recruited into the same NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DLG4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge