Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RET Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3385R-CY7
BOSSBS-3385R-CY7EA 929 EUR InStock
BOSSBS-3385R-CY7
Anti-RET Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The CD4 antigen is highly expressed on human T helper cells and thymocytes, and at lower levels on monocytes and dendritic cells. This integral membrane glycoprotein of approximately 58 kDa contains four external domains (D1 to D4) that show homology to members of the immunoglobulin (Ig) superfamily. It is responsible for the recognition of the MHC class II antigen. The CD4 antibody recognizes most thymocytes and about 65% of peripheral blood T cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RET
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge