Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SHC1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3412R-CY5.5
BOSSBS-3412R-CY5.5EA 929 EUR InStock
BOSSBS-3412R-CY5.5
Anti-SHC1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The SHC gene encodes for a signaling and transforming protein containing Src homology 2 and 3 (SH2 and SH3) domains. The SHC gene encodes 2 widely expressed overlapping proteins of 46 and 52 kD, both containing a C-terminal SH2 domain. Adjacent to the SH2 region is a glycine and proline rich region. These 2 proteins differ in their N terminals. SHC proteins are involved in mitogenic signal transduction and act by coupling growth factor receptors to the RAS signaling pathway. The protein encoded by the SHC1 gene is thought to act as an adaptor in many signal transduction pathways.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SHC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge