Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PEA15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3524R-FITC
BOSSBS-3524R-FITCEA 900 EUR InStock
BOSSBS-3524R-FITC
Anti-PEA15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
PED/PEA 15 (Phosphoprotein Enriched in Diabetes/Phosphoprotein Enriched in Astrocytes 15 kDa) is a widely expressed 15 kDa protein comprised of an N terminal region containing a canonical Death Effector Domain (DED) sequence and a nuclear export signal, and a C terminal region containing two serine phosphorylation sites. PED/PEA 15 has been implicated in the regulation of multiple cellular processes including apoptosis, integrin activation, and insulin sensitive glucose transport in insulin responsive cells. Phosphorylation of both serine 104 (a Protein Kinase C site) and serine 116 (a substrate of CaMKII and Akt) is required for PED/PEA 15 function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PEA15
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge