Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3542R-CY5.5
BOSSBS-3542R-CY5.5EA 900 EUR InStock
BOSSBS-3542R-CY5.5
Anti-PNK1/PNKP Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Mammalian PNK catalyzes the phosphorylation of DNA at 5'-hydroxyl termini and can dephosphorylate its 3'-phosphate termini. It plays an important function in DNA repair following ionizing radiation or oxidative damage.PNK has been reported to participate in the repair of DNA-double strand breaks via PARP-1-dependent nonhomologous end-joining.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PNK1/PNKP
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge