Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-DTNB Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3548R-CY5
BOSSBS-3548R-CY5EA 900 EUR InStock
BOSSBS-3548R-CY5
Anti-DTNB Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Dystrobrevin beta is a component of the dystrophin-associated protein complex and interacts with dystrophin short form DP71, syntrophins SNTG1 and SNTG2. It localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. 4 isoforms produced by alternative splicing have been identified.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DTNB
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge