Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HSPB8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3672R-A555
BOSSBS-3672R-A555EA 893 EUR InStock
BOSSBS-3672R-A555
Anti-HSPB8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Hsp22 (HSPB8) is a 196-amino acid protein that contains a central portion homologous to a highly conserved HSP-alpha crystallin domain common to all the small heat shock protein (HSP20) family members. Hsp22 is a monomeric protein which interacts with HSPB1. It displays temperature-dependent chaperone activity. The highest abundance of Hsp22 is in skeletal muscle, heart, and placenta. Mutations in the HSPB8 gene are associated with the inherited peripheral neuropathies, autosomal dominant distal hereditary motor neuropathy type IIA (dSMA) and axonal Charcot-Marie-Tooth disease type 2L (CMT2L).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSPB8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge