Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KLK11 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3799R-CY3
BOSSBS-3799R-CY3EA 900 EUR InStock
BOSSBS-3799R-CY3
Anti-KLK11 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Kallikrein 11 has two alternative splicing isoforms, known as the brain type and prostate type. Elevated serum levels of hK11 have been found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of this biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge