Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PLAA/Phospholipase A2 activator protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3807R-A750
BOSSBS-3807R-A750EA 874 EUR InStock
BOSSBS-3807R-A750
Anti-PLAA/Phospholipase A2 activator protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Phospholipase A2 activator protein (PLAP), activates PLA2 (phospholipase A2 enzyme) and is an important mediator of eicosanoid generation. Also known as a pro-inflammatory agent, PLAP has been found in inflamed tissues and synovial fluid from patients with rheumatoid arthritis. As such, it is believed to play an important role in the regulation of inflammatory diseases. The formation of PLAP can be stimulated by IL-1 beta and TNF-alpha.

Type: Primary
Antigen: PLAA/Phospholipase A2 activator protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/795
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge