Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NEDD8 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3812R-CY5
BOSSBS-3812R-CY5EA 900 EUR InStock
BOSSBS-3812R-CY5
Anti-NEDD8 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Ubiquitin like proteins fall into two classes: the first class, ubiquitin-like modifiers (UBLs) function as modifiers in a manner analogous to that of ubiquitin. Examples of UBLs are SUMO, Rub1 (also called Nedd8), Apg12 and Hub1. Proteins of the second class include Parkin, RAD23 and DSK2, are designated ubiquitin domain proteins (UDPs). These proteins contain domains that are related to ubiquitin but are otherwise unrelated to each other. In contrast to UBLs, UDPs are not proteolytically processed or conjugated to other proteins. Rub1 and the corresponding human homolog NEDD8 are activated by the E1 ubiquitin activating enzyme UBA2, that forms isopeptide linkages between thio esters. NEDD8 shows 80% homology to ubiquitin. The best known targets of Rub1 modification are members of the cullin family. Cullins are subunits of an E3 ubiquitin ligase complex called the Skp1 /Cul1/Cdc53 F box (SCF). The SCF promotes transfer of ubiquitin from a ubiquitin conjugating enzyme (E2) to the target protein. Rub1 modification may regulate SCF function or localization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEDD8
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge