Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MRPL39 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3835R-CY5.5
BOSSBS-3835R-CY5.5EA 900 EUR InStock
BOSSBS-3835R-CY5.5
Anti-MRPL39 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Two transcript variants encoding distinct isoforms have been described. A pseudogene corresponding to this gene is found on chromosome 5q.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MRPL39
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge