Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CACNA1E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3933R-FITC
BOSSBS-3933R-FITCEA 900 EUR InStock
BOSSBS-3933R-FITC
Anti-CACNA1E Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Calcium channels mediate the influx of calcium ions into the cell following membrane polarisation. R-type calcium channels such as Cav2.3 belong to the "high voltage-activated" group and are blocked by nickel. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Each of these proteins exists as multiple isoforms, either encoded by different genes or arising from alternative splicing of transcripts. Cav2.3 is an alpha-1 subunit and has 24 transmembrane segments, which form the pore through which ions pass into the cell. Calcium channels containing the Cav2.3 subunit may be involved in the modulation of firing patterns of neurons, which is important for information processing.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CACNA1E
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge