Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNB3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3943R-CY5.5
BOSSBS-3943R-CY5.5EA 900 EUR InStock
BOSSBS-3943R-CY5.5
Anti-GNB3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
G proteins are composed of 3 units, alpha, beta and gamma and are involved in the modulation and transduction of various transmembrane signalling systems. There are families of related genes for each subunit. GNB3 encodes a beta subunit. Beta subunits are required for GTPase activity, for replacement of GDP by GTP, regulate alpha subunits and for G protein-effector interactions. A single-nucleotide polymorphism (C825T) in this gene is associated with essential hypertension and obesity and is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein beta 3/GNB3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge