Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ENO3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-3979R-HRP
BOSSBS-3979R-HRPEA 900 EUR InStock
BOSSBS-3979R-HRP
Anti-ENO3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ENO3 is also known as beta enolase, one of the three enolase isoenzymes found in mammals. A switch from alpha enolase to beta enolase occurs in muscle tissue during development and ENO3, a homodimer, is found in skeletal muscle cells in the adult and appears to have a function in striated muscle development and regeneration. Mutations can result in decreased stability of the enzyme and be associated with a glycogen storage myopathy. This results in exercise-induced myalgias, generalized muscle weakness and fatigability.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ENO3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge