Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IRGM Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4015R-A680
BOSSBS-4015R-A680EA 890 EUR InStock
BOSSBS-4015R-A680
Anti-IRGM Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
IRGM belongs to a family of interferon-gamma inducible GTPases, belonging to the GTPase superfamily, and are selectively induced by IFN-gamma or bacterial lipopolysaccharide (LPS) stimulation. IRGM is primarily expressed in all cells derived from B-cell lineages, and is highly expressed in macrophages following IFN-gamma stimulation. IRGM has been shown to be required for host defences against a broad range of intracellular pathogens. Specifically, IRGM deficient mice show defects with intracellular microbial killing, phagosome maturation and autophagy. More recently IRGM has been shown to inhibit baseline hematopoietic proliferation and is required for a normal hematopoietic stem cell response to chemical and infectious stimuli.

Type: Primary
Antigen: IRGM
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/181
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge