Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ACADM Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4047R-CY3
BOSSBS-4047R-CY3EA 900 EUR InStock
BOSSBS-4047R-CY3
Anti-ACADM Rabbit Polyclonal Antibody (Cy3®)
Antikörper
ACADM protein is a medium chain specific (C4 to C12 straight chain) acyl Coenzyme A dehydrogenase. The enzyme catalyzes the initial step of the mitochondrial fatty acid beta oxidation pathway. ACADM expression is induced during periods of fasting, when reliance on fatty acids for energy is increased. Clinical phenotypes are associated with ACADM hereditary deficiency.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADM
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge