Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MAT1A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4054R-A555
BOSSBS-4054R-A555EA 876 EUR InStock
BOSSBS-4054R-A555
Anti-MAT1A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAT1A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge