Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SOX9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4121R-A488
BOSSBS-4121R-A488EA 893 EUR InStock
BOSSBS-4121R-A488
Anti-SOX9 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Exendin 4 is a 39 amino acid peptide found in venom from the Gila monster Helicoderma suspectum. It is a member of the glucagon secretin family of peptide hormones and neuropeptides. Exendin 4 is a potent agonist of the GLP1 receptor and hence a potent stimulator of insulin secretion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SOX9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge