Anti-MEK3 Thr222 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-4123R-A680
BOSSBS-4123R-A680EA 917 EUR InStock
BOSSBS-4123R-A680
Anti-MEK3 Thr222 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Dual specificity kinase. Is activated by cytokines and environmental stress in vivo. catalyses the concomitant phosphorylation of a threonine and a tyrosine residue in the MAP kinase p38.
Type: Primary
Antigen: MEK3 Thr222
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge