Anti-MAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-4128R-A555
BOSSBS-4128R-A555EA 893 EUR InStock
BOSSBS-4128R-A555
Anti-MAP4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Non-neuronal microtubule-associated protein. Promotes microtubule assembly.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MAP4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge