Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PKMYT1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4148R-CY3
BOSSBS-4148R-CY3EA 900 EUR InStock
BOSSBS-4148R-CY3
Anti-PKMYT1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
PKMYT1 is a member of the serine/threonine protein kinase family. It preferentially phosphorylates and inactivates cell division cycle 2 protein (cdc2), and thus acts as a negative regulator of entry into mitosis (G2 to M transition). It mediates phosphorylation of cdc2 predominantly on 'Thr-14' and is also involved in Golgi fragmentation. It may be involved in phosphorylation of cdc2 on 'Tyr-15' to a lesser degree, however tyrosine kinase activity is unclear and may be indirect. It may be a downstream target of Notch signaling pathway during eye development. PKMYT1 is negatively regulated by hyperphosphorylation during mitosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PKMYT1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge