Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-RPH3AL Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4183R-A555
BOSSBS-4183R-A555EA 893 EUR InStock
BOSSBS-4183R-A555
Anti-RPH3AL Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Rabphilin-3AL (rabphilin-3A-like), also known as RPH3AL or NOC2, is a cytoplasmic Rab GTPase effector. It contains one FYVE-type zinc finger and one Rab-binding (RBD) domain, but unlike its related protein, rabphilin-3A, rabphilin-3AL does not contain any C2 domains. Rabphilin-3AL is expressed in a variety of tissues, with highest levels found in kidney, skeletal muscle, pancreas, liver, ovary, stomach, heart and thyroid. It is believed to play a role regulating calcium-dependent secretory vesicle exocytosis in endocrine and exocrine cells. Via its RBD domain, rabphilin-3AL is capable of binding Rab 27a and, through this interaction, rabphilin-3AL is recruited to dense-core vesicles. With lower affinity, rabphilin-3AL can also bind Rab 3 and Rab 8 with its RBD domain. Through an interaction with Rab 3, rabphilin-3AL can inhibit G-protein signaling in endocrine pancreas and positively regulate insulin secretion. Rabphilin-3AL knockout mice display accumulation of secretory granules and irregular shape in exocrine cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPH3AL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge