Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-UGT2B4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4225R-HRP
BOSSBS-4225R-HRPEA 900 EUR InStock
BOSSBS-4225R-HRP
Anti-UGT2B4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of the UGT2B family are involved in the metabolism of bile acids, phenol derivatives, catecholestrogens and steroids. Although it is widely recognized that the liver is the major site of glucuronidation, it is now clear that UGT enzymes are also found in extra-hepatic tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: UGT2B4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge