Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDKN2C Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4270R-A680
BOSSBS-4270R-A680EA 890 EUR InStock
BOSSBS-4270R-A680
Anti-CDKN2C Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
p18 INK4C is a specific inhibitor of cdk4 / cdk6 (it interacts strongly with CDK6 and weakly with CDK4), and is expressed during G1 to S transition in the eukaryotic cell division cycle. It is maximally induced as cells enter S phase. Highest expression levels are found in skeletal muscle, but it is also found in the pancreas and heart.

Type: Primary
Antigen: CDKN2C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-168/168
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge