Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BPI Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4292R-CY5.5
BOSSBS-4292R-CY5.5EA 900 EUR InStock
BOSSBS-4292R-CY5.5
Anti-BPI Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
The antimicrobial protein Bactericidal Permeability Increasing protein (BPI) is a 55 kDa protein found in the primary granules of polymorhonuclear leukocytes (PMN). The cytotoxicity action of BPI is limited to gram negative bacteria, reflecting the high affinity of BPI for bacterial LPS. Binding of BPI to live bacteria via LPS causes anti-infective activites: 1) cytotoxicity via sequential damage to bacterial outer and inner lipid membranes, 2) neutralization of gram-negative bacterial LPS, 3) opsonization of bacteria to enhance phagocytosis by neutrophils.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge