Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CAMP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4735R-FITC
BOSSBS-4735R-FITCEA 900 EUR InStock
BOSSBS-4735R-FITC
Anti-CAMP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Cathelicidins are a family of antimicrobial proteins found in the peroxidase-negative granules of neutrophils. Along with the family of proteins known as defensins, cathelicidins participate in the first line of defense by preventing local infection and systemic invasion of microbes. FALL-39 precursor (FALL-39 peptide antibiotic, cationic anti-microbial protein, CAMP, CAP-18, HSD26) is a cathelicidin anti-microbial protein that contains the antibacterial peptide LL-37 (amino acids 134-170). In contrast to the defensins, which are cysteine-rich peptides that fold in ∫-pleated sheets, LL-37 is a cysteine-free peptide that can adopt an amphipathic å-helical conformation. LL-37 binds to bacterial lipopolysaccharides (LPS) and is a potent chemotactic factor for recruiting mast cells to sites of inflammation. LL-37 is present in inflammatory skin diseases that include psoriasis, sub-acute lupus erthematosus, dermatitis and nickel contact hypersensitivity. It is not found in normal skin epidermis. The secreted protein is expressed primarily in bone marrow, testis and neutrophils. The mouse and rat ortholog, CRAMP (cathelin-related antimicrobial peptide), is also part of the cathelicidin family of host defense peptides. These include precursors of potent antimicrobial peptides that direct antimicrobial activity against various microbial pathogens and also activate mesenchymal cells during wound repair. CRAMP is expressed in testis, spleen, stomach and intestine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAMP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge