Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NAPSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-4753R-HRP
BOSSBS-4753R-HRPEA 1350 EUR InStock
BOSSBS-4753R-HRP
Anti-NAPSA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Napsin A is an aspartic proteinase that belongs to the peptidase A1 family and plays a role in pneumocyte surfactant processing. It is a 420-amino acid polypeptide consisting of a 24-residue signal peptide, a 40-amino acid propart, the mature enzyme of 336 amino acids, and a C-terminal extension of 18 residues. The mature Napsin A protein contains 3 predicted disulfide bonds, 3 potential N-linked oligosaccharide attachment sites, an RGD motif, a recognition motif for integrin binding, in the C terminus, immediately before a 4-amino acid insert that is unique to aspartic proteinases. Highest levels of Napsin A have been detected in adult lung (type II pneumocytes), fetal lung, and kidney tissues. Napsin A is also expressed at lower levels in adult spleen and at very low levels in peripheral blood leukocytes. Human napsin A shares 72.6% sequence identity with the mouse homolog.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge