Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PFKFB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5004R-A647
BOSSBS-5004R-A647EA 893 EUR InStock
BOSSBS-5004R-A647
Anti-PFKFB1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
PFKFB1 belongs to the family of bifunctional 6-phosphofructo-2-kinase:fructose-2,6-biphosphatase enzymes. It forms a homodimer that is involved in both the synthesis and degradation of fructose 2,6-bisphosphate. The activity of this enzyme is thought to regulate glucose homeostasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PFKFB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge