Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TAAR8 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5060R-CY7
BOSSBS-5060R-CY7EA 900 EUR InStock
BOSSBS-5060R-CY7
Anti-TAAR8 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
FADS1 is a component of a lipid metabolic pathway that catalyzes biosynthesis of highly unsaturated fatty acids from precursor essential polyunsaturated fatty acids, linoleic acid and alphanlinolenic acid. It catalyzes the desaturation of dihomo gamma linoleic acid and eicosatetraenoic acid to generate arachidonic acid and eicosapentaenoic acid respectively.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAAR8
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge