Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PTPMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5103R-A680
BOSSBS-5103R-A680EA 890 EUR InStock
BOSSBS-5103R-A680
Anti-PTPMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This protein phosphatase specifically mediates the dephosphorylation of mitochondrial proteins and consequently plays a central role in ATP production. It probably has a preference for proteins phosphorylated on Ser and/or Thr residues compared to phosphorylation on Tyr residues. It is likely to be involved in the regulation of insulin secretion in pancreatic beta cells.

Type: Primary
Antigen: PTPMT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-201/201
Host: Rabbit
Isotype: IgG
Reactivity: Human; Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge