Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CREM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5135R-HRP
BOSSBS-5135R-HRPEA 900 EUR InStock
BOSSBS-5135R-HRP
Anti-CREM Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
CREM is a a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is derived from a multiexonic gene that encodes both activators and antagonists of cAMP-inducible transcription by differential splicing. Splice variants with antagonistic function lack 2 glutamine-rich domains and block cAMP-induced transcription, whereas an isoform that includes these glutamine-rich domains is a transcriptional activator.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CREM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge