Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-BID Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5231R
BOSSBS-5231REA 743 EUR InStock
BOSSBS-5231R
Anti-BID Rabbit Polyclonal Antibody
Antikörper
Bid, a BH3 domain containing proapoptotic Bcl2 family member, is localized in the cytosolic fraction of cells as an inactive precursor. Its active form is generated upon proteolytic cleavage by caspase 8 in the Fas signaling pathway. Cleaved Bid translocates to mitochondria and releases its potent proapoptotic activity, which in turn induces cytochrome c release and mitochondrial damage. The cytochrome c releasing activity of Bid was antagonized by Bcl2. Mutation in the SH3 domain can diminish the cytochrome c releasing activity. In animal model studies, Bid deficient mice are found resistant to the lethal effects of death factor signals relayed through Fas.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: BID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge