Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ESPL1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5304R-CY5.5
BOSSBS-5304R-CY5.5EA 930 EUR InStock
BOSSBS-5304R-CY5.5
Anti-ESPL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Separase is a cysteine protease that is essential for mitotic progression by separating sister chromatids. Each cell must receive one chromatid of every chromosome, during mitosis. Cohesin plays an important role in cohering sister chromatids during the prophase through anaphase stages of mitosis, making certain that genomic information is replicated accurately. As the cellular division process continues, separase destroys cohesin by means of cleavage, allowing the chromatids to separate and divide with the cell. Separase activity is highly regulated. It not only cleaves cohesin at the onset of anaphase but also cleaves itself, promoting downregulation of separase after anaphase. Should a human cell become an aneuploid (one too many or too few chromatids), the embryo most likely will not survive. Should the embryo survive, it will most likely develop severe birth defects or later develop malignant cancers.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ESPL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge