Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-EP300 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5339R-A488
BOSSBS-5339R-A488EA 922 EUR InStock
BOSSBS-5339R-A488
Anti-EP300 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
KAT3B/p300 is a transcriptional adapter protein (300 kDa)which is characterized by three cysteine and histidine rich regions and its C-terminus specifically binds the adenovirus E1A protein. KAT3B and associated proteins are components of TATA-binding protein (TBP)complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265 kDa nuclear protein designated KAT3A/CBP(for CREB-binding protein). KAT3B and KAT3A are homologous to each other.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EP300
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge