Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-PMVK Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5371R-A750
BOSSBS-5371R-A750EA 890 EUR InStock
BOSSBS-5371R-A750
Anti-PMVK Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PMVK is a 192 amino acid peroxisomal enzyme belonging to the nucleoside monophosphate (NMP) kinase family and is expressed in heart, liver, skeletal muscle, kidney, and pancreas with lower expression in brain, placenta and lung. Induced by sterol, PMVK participates in isopentenyl diphosphate biosynthesis via the mevalonate pathway. PMVK catalyses the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate in the fifth reaction of the cholesterol biosynthetic pathway. PMVK exists as a monomer and is encoded by a gene located on human chromosome 1, which houses over 3,000 genes and is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome.

Type: Primary
Antigen: PMVK
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 15-120/192
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge