Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GIT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5372R-FITC
BOSSBS-5372R-FITCEA 1350 EUR InStock
BOSSBS-5372R-FITC
Anti-GIT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Heterotrimeric G protein-mediated signal transduction is a dynamically regulated process with the intensity of signal decreasing over time despite the continued presence of the agonist (1,2). G protein-coupled receptor kinases (GRKs) are activated by activated G protein-coupled receptors, and they function to phosphorylate and inactivate cell surface receptors in the heterotrimeric G protein signaling cascade (3,4). GIT1 (for GRK-interactor 1) and GIT2 are GTPase-activating proteins (GAP) for members of the ADP ribosylation factor (ARF) family of small GTP-binding proteins, which are involved in vesicular trafficking (5,6). GIT1 overexpression results in reduced internalization and resensitization of b2-adrenergic receptor, thus reducing b2-adrenergic receptor signaling (5).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge