Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HDAC1 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5387R
BOSSBS-5387REA 1090 EUR InStock
BOSSBS-5387R
Anti-HDAC1 Rabbit Polyclonal Antibody
Antikörper
Reversible acetylation of highly conserved lysine residues within the N-terminal tail domains of core histones, plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone acetylation is a dynamic process determined by the net activities of histone acetyltransferases (HATs) and the competing enzymes histone deacetylases (HDACs). Histone deacetylases activities are often, but not always, associated with transcriptional repression and nucleosomal condensations. Recruitment of the multiprotein complexes to promoter sites occurs by many sequence specific DNA-binding proteins such as unliganded nuclear hormone receptors, DP1-E2F, YY1 and Rb family of transcription factors, transcriptional repressors and tumor suppressors (e.g. BRCA1). Aberrant recruitment of HDACs by certain oncoproteins may occur in certain neoplastic diseases. Belongs to the histone deacetylase family.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge