Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MAP4K4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5491R-FITC
BOSSBS-5491R-FITCEA 930 EUR InStock
BOSSBS-5491R-FITC
Anti-MAP4K4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
MAP4K4 (Mitogen-activated protein kinase kinase kinase kinase 4) belongs to the serine/threonine protein kinase family and specifically activates MAPK8/JNK. It may play a role in the response to environmental stress and is thought to function through the MAP3K7-MAP2K4-MAP2K7 kinase cascade, and mediate the TNF alpha signaling pathway. It interacts with the SH3 domain of the adapter protein Nck. MAP4K4-dependent signaling inhibits PPARG responsive gene expression, adipogenesis, and insulin stimulated glucose transport.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAP4K4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge