Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-NDEL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5523R-FITC
BOSSBS-5523R-FITCEA 930 EUR InStock
BOSSBS-5523R-FITC
Anti-NDEL1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Nudel is important for normal cortical development. It is invovled in microtubule organization, nuclear translocation, and neuronal positioning in concert with various other factors (including Lis1, Pafah1b1, Pahfah1b2, dynein, dynorphin A and cdk5). Western blot analysis of mouse tissues shows abundant expression of Nudel in brain and testis, and much lower expression in heart, liver, kidney, and skeletal muscle. In fractionated rat brain, Nudel and Lis1 are both found in fractions enriched for postsynaptic density proteins. Immunostaining of embryonic day 18 mouse brain sections revealed staining of migrating neurons and thalamocortical axons of the intermediate zone of the developing cerebral cortex, as well as several other developing brain regions. The deduced protein contains 345 amino acids and has a calculated molecular mass of 38.4 kDa. It has a coiled coil motif (residues 19 to 201), followed by several potential phosphorylation sites for casein kinase II, protein kinase C or CDK5. Nudel shares about 50% identity with mouse and human NUDE proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NDEL1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge