Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5626R-HRP
BOSSBS-5626R-HRPEA 1350 EUR InStock
BOSSBS-5626R-HRP
Anti-SNCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The synucleins, including Alpha-synuclein (also designated NACP for nonamyloid component precursor),Beta-synuclein (also designated PNP 14 for neuroprotein 14)and Gamma-synuclein (also designated persyn or BCSG1 for breast cancer-specific gene 1)are presynaptic protein abundant in neurons. Alpha-synuclein, a component of Alzheimer’s disease amyloid plaques, is localized to neuronal cell bodies and synapses. Coordinate expression of Alpha-synucleinand Beta-synuclein may be important during hematopoetic cell differentiation. In patients with Parkinson’s disease, a mutant form of Alpha-synuclein has been found and Gamma-synuclein is associated with axonal pathology.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge