Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SNCA phospho Ser129 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5628R-A750
BOSSBS-5628R-A750EA 921 EUR InStock
BOSSBS-5628R-A750
Anti-SNCA phospho Ser129 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The synucleins, including Alpha-synuclein (also designated NACP for nonamyloid component precursor),Beta-synuclein (also designated PNP 14 for neuroprotein 14)and Gamma-synuclein (also designated persyn or BCSG1 for breast cancer-specific gene 1)are presynaptic protein abundant in neurons. Alpha-synuclein, a component of Alzheimer?s disease amyloid plaques, is localised to neuronal cell bodies and synapses. Coordinate expression of Alpha-synucleinand Beta-synuclein may be important during hematopoetic cell differentiation. In patients with Parkinson?s disease, a mutant form of Alpha-synuclein has been found and Gamma-synuclein is associated with axonal pathology.

Type: Primary
Antigen: Alpha synuclein Ser129
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge