Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SYT6 Thr418 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-5636R-A750
BOSSBS-5636R-A750EA 1350 EUR InStock
BOSSBS-5636R-A750
Anti-SYT6 Thr418 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Synaptotagmins (Syts) are a large gene family of synaptic vesicle integral membrane proteins. Syt functions as regulators of both exocytosis /endocytosis and is involved in neurotransmitter secretion from small secretory vesicles. 13 isoforms of Syt have been identified(SytI-XIII). SynaptotagminVI is localised to the endoplasmic reticulum and/or Golgi like perinuclear compartment. It may be important for trafficking and calcium signaling as it is specially expressed in nonneuronal tissues. Also SytVI is present in the acrosomal region of mammalian spermatozoa. The cytosolic domain of SytVI can abrogate exocytosis by competing with the endogenous protein for essential interactions with the fusion machinery involved in a acrosomal exocytosis.

Type: Primary
Antigen: SYT6 Thr418
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge